Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytosolic 5'-nucleotidase 1B Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE1005
Description
This Anti-Cytosolic 5′-nucleotidase 1B Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Cytosolic 5′-nucleotidase 1B.
Specifications
Cytosolic 5'-nucleotidase 1B | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NT5C1B; AIRP; FKSG85 | |
BCP = BLUE CARRIER PROTEIN-conjugated linear peptide corresponding to human Cytosolic 5'-nucleotidase 1B. | |
100 μg | |
NP_001002006 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction