Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Serine Rabbit anti-All, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5917
Description
Anti-D-Serine Antibody is an antibody against D-Serine for use in ELISA.
Specifications
D-Serine | |
Polyclonal | |
Liquid rabbit serum with 0.05% sodium azide. | |
D-Serine-Glutaraldehyde-BSA | |
Primary | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
ELISA | |
Unconjugated | |
Rabbit | |
100 μL | |
All species | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction