Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dishevelled associated activator of morphogenesis 1, Mouse, Clone: 4H3, Abnova™

Catalog No. 89001220 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-220 Supplier Abnova Corporation Supplier No. H00023002M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant DAAM1.

Functions of the cell cortex, including motility, adhesion, and cytokinesis, are mediated by the reorganization of the actin cytoskeleton and recent evidence suggests a role for the Formin homology (FH) proteins in these processes. The protein encoded by this gene contains FH domains and belongs to a novel FH protein subfamily implicated in cell polarity. Wnt/Fz signaling activates the small GTPase Rho, a key regulator of cytoskeleton architecture, to control cell polarity and movement during development. Activation requires Dvl-Rho complex formation, an assembly mediated by this gene product, which is thought to function as a scaffolding protein. Evidence of alternative splicing has been observed for this gene but the full-length nature of these variants has not been determined. [provided by RefSeq

Sequence: MAPRKRGGRGISFIFCCFRNNDHPEITYRLRNDSNFALQTMEPALPMPPVEELDVMFSELVDELDLTDKHREAMFALPAEKKWQIYCSKKKDQEENKGATSWPEFYIDQL

Specifications

Antigen dishevelled associated activator of morphogenesis 1
Applications ELISA, In situ PLA, Western Blot
Classification Monoclonal
Clone 4H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant DAAM1.
Formulation PBS with no preservative; pH 7.4
Gene DAAM1
Gene Accession No. NM_014992
Gene Alias FLJ41657/KIAA0666
Gene Symbols DAAM1
Host Species Mouse
Immunogen DAAM1 (NP_055807, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23002
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.