Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAAM1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512292
Description
This antibody is predicted to react with mouse based on sequence homology.
Functions of the cell cortex, including motility, adhesion, and cytokinesis, are mediated by the reorganization of the actin cytoskeleton. Recent evidence suggests a role for the Formin homology (FH) proteins in these processes. DAAM1 contains FH domains and belongs to a novel FH protein subfamily implicated in cell polarity. Wnt/Fz signaling activates the small GTPase Rho, a key regulator of cytoskeleton architecture, to control cell polarity and movement during development. Activation requires Dvl-Rho complex formation, an assembly mediated by DAAM1, which is thought to function as a scaffolding protein.
Specifications
DAAM1 | |
Polyclonal | |
DAAM1 | |
Disheveled-associated activator of morphogenesis 1;DAAM1;KIAA0666 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23002 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9Y4D1 | |
DAAM1 | |
KLH conjugated synthetic peptide between 45-74 amino acids from the N-terminal region of human DAAM1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction