Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB2, Mouse, Clone: 3D6, Abnova™

Catalog No. 89021250 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-250 Supplier Abnova Corporation Supplier No. H00001601M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DAB2.

DAB2 mRNA is expressed in normal ovarian epithelial cells but is down-regulated or absent from ovarian carcinoma cell lines. The 770-amino acid predicted protein has an overall 83% identity with the mouse p96 protein, a putative mitogen-responsive phosphoprotein; homology is strongest in the amino-terminal end of the protein in a region corresponding to the phosphotyrosine interaction domain. The down-regulation of DAB2 may play an important role in ovarian carcinogenesis. This gene was initially named DOC2 (for Differentially expressed in Ovarian Cancer) and is distinct from the DOC2A and DOC2B genes (for double C2-like domains, alpha and beta). [provided by RefSeq

Sequence: MSNEVETSATNGQPDQQAAPKAPSKKEKKKGPEKTDEYLLARFKGDGVKYKAKLIGIDDVPDARGDKMSQDSMMKLKGMAAAGRSQGQHKQRIWVNISLSGIKIIDEKTGVIEHEHPVNKISFIARDVTDNRAFGYVCGGEGQHQFFAIKTGQQAEPLVVDLKDLFQVIYNVKKKEEEKKKIEEASKAVENGSEALMILDDQTNKLKSGVDQMDLFGDMSTPPDLNSPTESKDILLVDLNSEIDTNQNSLRENPFLTNGITSCSLPRPTPQASFLPENAFSANLNFFPTPNPDPFRDDPFTQPDQSTPSSFDSLKSPDQKKENSSSSSTPLSNGPLNGDVDYFGQQFDQISNRTGKQEAQAGPWPFSSSQTQPAVRTQNGVSEREQNGFSVKSSPNPFVGSPPKGLSIQNGVKQDLESSVQSSPHDSIAIIPPPQSTKPGRGRRTAKSSANDLLASDIFAPPVSEPSGQASPTGQPTALQPNPLDLFKTSAPAPVGPLVGLGGVTVTLPQAGPWNTASLVFNQSPSMAPGAMMGGQPSGFSQPVIFGTSPAVSGWNQPSPFAASTPPPVPVVWGPSASVAPNAWSTTSPLGNPFQSNIFPAPAVSTQPPSMHSSLLVTPPQPPPRAGPPKDISSDAFTALDPLGDKEIKDVKEMFKDFQLWQPPAVPARKGEQTSSGTLSAFASYFNSKVGIPQENADHDDFDANQLLNKINEPPKPAPRQVSLPVTKSTDNAFENPFFKDSFGSSQASVASSQPVSSEMYRDPFGNPFA

Specifications

Antigen DAB2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DAB2.
Formulation PBS with no preservative; pH 7.4
Gene DAB2
Gene Accession No. BC003064
Gene Alias DOC-2/DOC2/FLJ26626
Gene Symbols DAB2
Host Species Mouse
Immunogen DAB2 (AAH03064, 1 a.a. ∼ 770 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1601
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.