Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP, Mouse, Clone: 3C5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DAP.
Supplier: Abnova Corporation H00001611M01
Description
This gene encodes a basic, proline-rich, 15kDa protein. The protein acts as a positive mediator of programmed cell death that is induced by interferon-gamma. [provided by RefSeq
Sequence: MSSPPEGKLETKAGHPPAVKAGGMRIVQKHPHTGDTKEEKDKDDQEWESPSPPKPTVFISGVIARGDKDFPPAAAQVAHQKPHASMDKHPSPRTQHIQQPRKSpecifications
DAP | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC002726 | |
DAP | |
DAP (AAH02726, 1 a.a. ∼ 102 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
3C5 | |
Mouse monoclonal antibody raised against a full-length recombinant DAP. | |
DAP | |
MGC99796 | |
Mouse | |
Affinity chromatography | |
RUO | |
1611 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction