Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP Kinase Mouse anti-Human, Unlabeled, Clone: 17, BD

Catalog No. BDB610291 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610291 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610291 Supplier BD Biosciences Supplier No. 610291
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chronic exposure to extracellular signals such as interferons induce the inhibition of cell proliferation followed by cell death. The gene for DAP Kinase (Death Associated Protein Kinase) was identified using a novel approach named Technical Knockout. Briefly, Hela cells were transfected with an antisense cDNA expression library, then exposed to Interferon-γ. The surviving cells, with their antisense cDNAs, were rescued and the “protecting” genes isolated. The DAP Kinase gene encodes a protein of 1423 amino acids, a molecular weight of 160kDa, a kinase domain at its amino terminal region, ankyrin repeats in the middle region, and a death domain at the extreme C-terminus. DAP Kinase phosphorylates at Ser/Thr residues in a Ca2+/Calmodulin-dependent fashion. It has been demonstrated that Ca2+/Calmodulin binds directly to DAP Kinase at its amino terminal region. In addition, immunostaining studies localized DAP Kinase in association with the actin filaments where it may phosphorylate myosin light chain. Thus, this novel cytoskeletal and Ca2+/Calmodulin-dependent protein kinase plays a role in interferon-γ-induced cell death.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DAP Kinase
Applications Western Blot
Classification Monoclonal
Clone 17
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.