Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAP12 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB4070
Description
Anti-DAP12 Antibody detects level of DAP12 and has been published and validated for use in immunoprecipitation.
Specifications
| DAP12 | |
| Polyclonal | |
| Purified immunoglobulin. Liquid in phosphate buffered saline with 0.08% sodium azide. | |
| Rabbit | |
| 100 μg | |
| NM_003332.2; NM_198125.1 | |
| −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
| Immunoprecipitation | |
| Unconjugated | |
| TYROBP; DAP12; PLO-SL; KARAP; PLOSL | |
| Recombinant DAP12 protein. | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction