Learn More
DARC, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DARC peptide using ARM Technology.
Supplier: Abnova Corporation H00002532K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a glycosylated membrane protein and a nonspecific receptor for several chemokines. The encoded protein is the receptor for the human malarial parasites Plasmodium vivax and Plasmodium knowlesi. Polymorphisms in this gene are the basis of the Duffy blood group system. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DARC | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DARC | |
DARC | |
A synthetic peptide of human DARC | |
RUO | |
2532 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
Duffy blood group, chemokine receptor | |
DARC | |
CCBP1, CD234, Dfy, FY, GPD, GpFy, WBCQ1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.