Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dardarin Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9682MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9682MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9682MI Supplier MilliporeSigma Supplier No. AB9682
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Dardarin Antibody detects level of Dardarin and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dardarin
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit Serum. Liquid.
Gene Symbols LRRK2; AURA17; RIPK7; ROCO2; dardarin; FLJ45829; PARK8; DKFZp434H2111; Dardarin; EC 2.7.11.1
Host Species Rabbit
Immunogen Synthetic peptide from rat LRRK2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_198578.2
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.