Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARPP-32 Mouse, Unlabeled, Clone: 15, BD

Catalog No. BDB611520 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611520 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611520 Supplier BD Biosciences Supplier No. 611520
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dopaminergic signaling in midbrain neurons is essential to multiple brain functions and involves the activation of dopamine receptors, such as D1 and D2, which regulate the phosphorylation state of DARPP-32 (dopamine and cyclic AMP-regulated phospho-protein of Mr = 32,000). D1 receptor ligation causes activation of PKA and phosphorylation of DARPP-32 at Thr-34, which converts DARPP-32 to a potent inhibitor of protein phosphatase 1 (PP1). In addition, DARPP-32 is converted to an inhibitor of PKA via phosphorylation at Thr-75 by
cyclin-dependent kinase 5 (Cdk5). D2 receptor ligation inhibits PKA and activates protein phosphatase 2B/calcineurin causing dephosphorylation of DARPP-32. The major function of DARPP-32 may be to inhibit the activity of PP1, which controls the phosphorylation state of neurotransmitter receptors, ion channels, ion pumps, and transcription factors. DARPP-32 -/- mice are defective in the physiologicial and behavioral responses to dopamine. Thus, DARPP-32, a bifunctional signal transduction molecule that differentially controls a Ser/Thr kinase and a Ser/Thr phosphatase, is a critical element of dopaminergic neurotransmission and normal brain function.

Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human DARPP-32 aa. 70-181
Formula Weight [Chemical]: 32kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DARPP-32
Applications Western Blot
Classification Monoclonal
Clone 15
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human DARPP-32 aa. 70-181
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.