Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DARPP-32 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB9208
Description
Detect DARPP-32 using this Anti-DARPP-32 Antibody, phosphoThr75 validated for use in western blotting.
Specifications
DARPP-32, phosphoThr75 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in 10mM HEPES, pH 7.5, 150mM NaCl, 100 μg per mL BSA with 50% glycerol. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Western Blot | |
Unconjugated | |
PPP1R1B; DARPP32; DARPP-32; FLJ20940 | |
Synthetic peptide from human/mouse DARPP-32, phosphoThr75. | |
100 μL | |
NM_032192.2; NM_181505.2 | |
−20°C to −80°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction