Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAX-1 Mouse anti-Human, Clone: 2F4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABD398
Description
This Anti-DAX-1, clone 2F4 Antibody is validated for use in Western Blotting and Immunocytochemistry and Immunohistochemistry for the detection of DAX-1.
Specifications
DAX-1 | |
Monoclonal | |
Unconjugated | |
NP_000466 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
2F4 | |
Mouse monoclonal IgG1κ Ascites with 0.05% sodium azide. | |
NR0B1, DAX1, AHC | |
Linear peptide corresponding to Human DAX-1 (amino acids 135-166). | |
100 μL | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction