Learn More
DAZL, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DAZL peptide using ARM Technology.
Supplier: Abnova Corporation H00001618K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The DAZ (Deleted in AZoospermia) gene family encodes potential RNA binding proteins that are expressed in prenatal and postnatal germ cells of males and females. The protein encoded by this gene is localized to the nucleus and cytoplasm of fetal germ cells and to the cytoplasm of developing oocytes. In the testis, this protein is localized to the nucleus of spermatogonia but relocates to the cytoplasm during meiosis where it persists in spermatids and spermatozoa. Transposition and amplification of this autosomal gene during primate evolution gave rise to the DAZ gene cluster on the Y chromosome. Mutations in this gene have been linked to severe spermatogenic failure and infertility in males. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DAZL | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DAZL | |
DAZL | |
A synthetic peptide of human DAZL | |
RUO | |
1618 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
deleted in azoospermia-like | |
DAZL | |
DAZH, DAZL1, DAZLA, MGC26406, SPGYLA | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.