Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBF4 Mouse anti-Human, Clone: 7H8.1, MilliporeSigma™

Catalog No. MABE338MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE338MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE338MI Supplier MilliporeSigma Supplier No. MABE338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-DBF4 antibody, clone 7H8.1 detects level of DBF4 and has been published and validated for use in DBF4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DBF4
Classification Monoclonal
Clone 7H8.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_006707
Gene Symbols DBF4, ASK, DBF4A, ZDBF1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human DBF4 near the C-terminus.
Purification Method Protein G purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.