Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBN1, Mouse, Clone: 2E11, Abnova™

Catalog No. 89021261 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-261 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-261 Supplier Abnova Corporation Supplier No. H00001627M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant DBN1.

The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene. [provided by RefSeq]

Sequence: MAGVSFSGHRLELLAAYEEVIREESAADWALYTYEDGSDDLKLAASGEGGLQELSGHFENQKVMYGFCSVKDSQAALPKYVLINWVGEDVPDARKCACASHVAKVAEFFQGVDVIVNASSVEDIDAGAIGQRLSNGLARLSSPVLHRLRLREDENAEPVGTTYQKTDAAVEMKRINREQFWEQAKKEEELRKEEERKKALDERLRFEQERMEQERQEQEERERRYREREQQIEEHRRKQQTLEAEEAKRRLKEQSIFGDHRDEEEETHMKKSESEVEEAAAIIAQRPDNPREFFKQQERVASASAGSCDVPSPFNHRPGSHLDSHRRMAPTPIPTRSPSDSSTASTPVAEQIERALDEVTSSQPPPLPPPPPPAQETQEPSPILDSEETRAAAPQAWAGPMEEPPQAQAPPRGPGSPAEDLMFMESAEQAVLAAPVEPATADATEVHDAADTIETDTATADTTVANNVPPAATSLIDLWPGNGEGASTLQGEPRAPTPPSGTEVTLAEVPLLDEVAPEPLLPAGEGCATLLNFDELPEPPATFCDPEEVEGEPLAAPQTPTLPSALEELEQEQEPEPHLLTNGETTQKEGTQASEGYFSQSQEEEFAQSEELCAKAPPPVFYNKPPEIDITCWDADPVPEEEEGFEGGD

Specifications

Antigen DBN1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant DBN1.
Formulation PBS with no preservative; pH 7.4
Gene DBN1
Gene Accession No. BC000283
Gene Alias D0S117E/DKFZp434D064
Gene Symbols DBN1
Host Species Mouse
Immunogen DBN1 (AAH00283, 1 a.a. ∼ 649 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1627
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.