Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN Rat anti-Mouse, Clone: BMD30, MilliporeSigma™

Catalog No. MABF99MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF99MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF99MI Supplier MilliporeSigma Supplier No. MABF99

Rat Monoclonal Antibody

This Anti-DC-SIGN Antibody, clone BMD30 is validated for use in immunohistochemistry for the detection of DC-SIGN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DC-SIGN
Applications Immunohistochemistry
Classification Monoclonal
Clone BMD30
Conjugate Unconjugated
Formulation Purified Rat Monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_573501
Gene Symbols Cd209a, Cire
Host Species Rat
Immunogen Histidine-tagged recombinant protein corresponding to mouse DC-SIGN.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.