Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514046
Description
Doublecortin-like kinase (DCAMKL1)(Ser/Thr protein kinase family) is essential for proper neurogenesis, neuronal migration, and axonal wiring. DCAMKL1 is involved in a calcium-signaling pathway controling neuronal migration in the developing brain, and participates in functions of the mature nervous system. DCAMKL1 protein shares high homology with doublecortin (DCX). DCLK, but not DCX, is highly expressed in regions of active neurogenesis in the neocortex and cerebellum. DCAMKL1 controls mitotic division by regulating spindle formation and also determines the fate of neural progenitors during cortical neurogenesis.
Specifications
DCLK1 | |
Polyclonal | |
Unconjugated | |
O15075, Q9JLM8 | |
DCLK1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human DCAMKL1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
0.5 mg/mL | |
DCLK1 | |
Serine/threonine-protein kinase DCAMKL1, Doublecortin-like and CAM kinase-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13175, 9201 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction