Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCC Mouse anti-Human, Unlabeled, Clone: G97-449, BD

Catalog No. BDB554223 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554223 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554223 Supplier BD Biosciences Supplier No. 554223
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

One of the most common regions of allelic loss in colorectal tumors is chromosome 18, which is lost in more than 70% of carcinomas, and in almost 50% of late adenomas. This region of loss has been mapped to chromosome 18q and a gene called Deleted in Colorectal Cancer (DCC). DCC encodes an ∽185kDa glycoprotein with significant homology to the neural cell adhesion molecule and other related cell surface glycoproteins. The predicted amino acid sequence of DCC cDNA consists of a 1448 amino acid (aa) long transmembrane phosphoprotein. The extracellular domain consists of 1098 amino acids and has 42% sequence homology to cell adhesion proteins of the neural cell adhesion molecule (N-CAM) family. DCC mRNA is found to be expressed in normal colonic mucosa, but its expression is reduced or absent in the majority of colorectal carcinomas. The loss of heterozygosity and subsequent alteration of DCC expression has also been observed in tumors of non-colorectal origin. Clone G97-449 recognizes human DCC. A truncated recombinant protein containing the intracellular domain of the human DCC was used as immunogen.

Host Species: Mouse
Clone: G97-449
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human DCC
Formula Weight [Chemical]: 175-190kDa

Flow Cytometry, Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCC
Applications Western Blot
Classification Monoclonal
Clone G97-449
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human DCC
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.