Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCIR Mouse anti-Human, Alexa Fluor 647, Clone: I3-612, BD

Catalog No. BDB558220 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558220 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558220 Supplier BD Biosciences Supplier No. 558220
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The I3-612 antibody reacts with DCIR, or dendritic cell immunoreceptor, a newly identified member of the calcium-dependent (C-type) lectin family. DCIR is a type II membrane glycoprotein with a single carbohydrate recognition domain (CRD). Its cytoplasmic tail contains a consensus immunoreceptor tyrosine-based inhibitory motif (ITIM). It is expressed on monocytes, macrophages, B-lymphocytes, granulocytes, but not on NK or T cells. DCIR is strongly expressed on dendritic cells derived from peripheral blood monocytes cultured in presence of GM-CSF and IL-4. Down-regulation is observed on these with further maturation using TNF-α or LPS. Reports describe the detection of two distinct inhibitory signals depending on its ITIM tyrosine residue.

Host Species: Mouse
Clone: I3-612
Isotype: IgG1, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen DCIR
Applications Flow Cytometry
Classification Monoclonal
Clone I3-612
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.