Learn More
DCLRE1B, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DCLRE1B peptide using ARM Technology.
Supplier: Abnova Corporation H00064858K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links (Dronkert et al., 2000 [PubMed 10848582]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DCLRE1B | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DCLRE1B | |
DCLRE1B | |
A synthetic peptide of human DCLRE1B | |
RUO | |
64858 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae) | |
DCLRE1B | |
APOLLO, DKFZp686C24187, FLJ12810, FLJ13998, SNM1B | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.