Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLRE1B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525341
Description
DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links.
Specifications
DCLRE1B | |
Polyclonal | |
DCLRE1B | |
APOLLO, SNM1B, SNMIB, 5′ exonuclease Apollo, DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae), PSO2 homolog, SNM1 homolog B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140917, 64858 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9H816, Q8C7W7 | |
DCLRE1B | |
KLH conjugated synthetic peptide between 208-236 amino acids from the central region of human DCLRE1B | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction