Learn More
DCP1A, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DCP1A peptide using ARM Technology.
Supplier: Abnova Corporation H00055802K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Decapping is a key step in general and regulated mRNA decay. The protein encoded by this gene is a decapping enzyme. This protein and another decapping enzyme form a decapping complex, which interacts with the nonsense-mediated decay factor hUpf1 and may be recruited to mRNAs containing premature termination codons. This protein also participates in the TGF-beta signaling pathway. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| DCP1A | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| DCP1A | |
| DCP1A | |
| A synthetic peptide of human DCP1A | |
| RUO | |
| 55802 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| DCP1 decapping enzyme homolog A (S. cerevisiae) | |
| DCP1A | |
| FLJ21691, HSA275986, Nbla00360, SMAD4IP1, SMIF | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.