Learn More
CD263 (TRAIL-R3) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA1956
Description
PA1-956 detects decoy receptor 1 (DcR1) from human and rat tissues and cells. PA1-956 has been successfully used in FACS and Western blot procedures. By Western blot, this antibody detects an ∽30 kDa protein representing DcR1 from HEK293 cell lysate. PA1-956 immunizing peptide corresponds to amino acid residues 24-35 of human DcR1. PA1-956 immunizing peptide (Cat. No. PEP-047) is available for use in neutralization and control experiments.
DcR1 / TRAIL-R3 / TRID / LIT is one of the two putative decoy receptors identified for TRAIL, one of the members of TNF family of apoptosis inducing proteins. The other putative decoy receptor is called DcR2 / TRUNDD / TRAIL-R4. DcR1 is attached to the cell surface through glycophospholipid anchor. It has the extracellular TRAIL binding domain but lacks the cytoplasmic domain to induce apoptotic signal. Hence overexpression of DcR1 inhibits the TRAIL induced apoptosis.
Specifications
CD263 (TRAIL-R3) | |
Polyclonal | |
Unconjugated | |
O14798, P50591 | |
TNFRSF10C, TNFSF10 | |
Synthetic Peptide: Y(24) S A T T A R Q E E V P(35) C | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
1 mg/mL | |
TNFSF10 | |
CD263, DCR1, LIT, MGC149501, MGC149502, TRAILR3, TRID | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
246775, 8743, 8794 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.