Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DcR2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB16943
Description
Anti-DcR2 Antibody, intracellular domain detects level of DcR2 and has been published and validated for use in western blot, immunohistochemistry.
Specifications
DcR2 | |
Polyclonal | |
Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide. | |
Rabbit | |
100 μg | |
NM_003840.3 | |
2-8°C in undiluted aliquots for up to 12 months |
Immunohistochemistry, Western Blot | |
Unconjugated | |
TNFRSF10D; CD264; DcR2; TRUNDD; TRAIL-R4; TRAILR4; DCR2 | |
Polyclonal antibody raised against a peptide corresponding to amino acids 249-263 of human DcR2 precursor (Marsters et al. 1997; Degli-Esposti et al. 1997; Pan et al. 1997). | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction