Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DDB1 Mouse, Unlabeled, Clone: 8, BD

Catalog No. BDB612488 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612488 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612488 Supplier BD Biosciences Supplier No. 612488

Mouse Monoclonal Antibody

DNA lesions caused by chemical mutagens or radiation are corrected via nucleotide excision repair (NER). The NER system includes multiple proteins involved in xeroderma pigmentosum (XP) disorders, a pathology that causes hypersensitivity to sunlight and higher incidence of skin cancer. The proteins that cause these disorders are XP proteins and include XPA, XPB, XPC, XPD, XPF, and XPG. There are six repair complexes in the NER system composed of 15-18 proteins that include XPA, XPC, XPF, TFIIH, and hHR23B. In addition to these proteins, UV damage DNA-binding (UV-DDB) protein activity has also been associated with the NER system due to the fact that UV-DDB activity is absent in a subset of XPE Ddb- patients. UV-DDB consists of two subunits, DDB1 and DDB2, which can be injected into XPE cells to restore DNA repair synthesis. UV-DDB activity may be involved in the early stages of NER when it may promote recognition of the damaged DNA through DDB2. In addition, DDB1 can bind the histone acetyltransferase p300, which may be important for chromatin remodeling during the early stages of NER. Thus, UV-DDB activity may be important for recognition of specific types of DNA damage during NER.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DDB1
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human DDB1 aa. 739-935
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.