Learn More
dopa decarboxylase (aromatic L-amino acid decarboxylase) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DDC protein.
Supplier: Abnova Corporation H00001644B01
Description
The encoded protein catalyzes the decarboxylation of L-3,4-dihydroxyphenylalanine (DOPA) to dopamine, L-5-hydroxytryptophan to serotonin and L-tryptophan to tryptamine. Defects in this gene are the cause of aromatic L-amino-acid decarboxylase deficiency (AADCD). AADCD deficiency is an inborn error in neurotransmitter metabolism that leads to combined serotonin and catecholamine deficiency. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Sequence: MNASEFRRRGKEMVDYVANYMEGIEGRQVYPDVEPGYLRPLIPAAAPQEPDTFEDIINDVEKIIMPGVTHWHSPYFFAYFPTASSYPAMLADMLCGAIGCIGFSWAASPACTELETVMMDWLGKMLELPKAFLNEKAGEGGGVIQGSASEATLVALLAARTKVIHRLQAASPELTQAAIMEKLVAYSSDQAHSSVERAGLIGGVKLKAIPSDGNFAMRASALQEALERDKAAGLIPFFMVATLGTTTCCSFDNLLEVGPICNKEDIWLHVDAAYAGSAFICPEFRHLLNGVEFADSFNFNPHKWLLVNFDCSAMWVKKRTDLTGAFRLDPTYLKHSHQDSGLITDYRHWQIPLGRRFRSLKMWFVFRMYGVKGLQAYIRKHVQLSHEFESLVRQDPRFEICVEVILGLVCFRLKGSNKVNEALLQRINSAKKIHLVPCHLRDKFVLRFAICSRTVESAHVQRAWEHIKELAADVLRAERESpecifications
DOPA Decarboxylase | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human DDC protein. | |
DDC | |
AADC | |
Mouse | |
50 μL | |
Yes | |
1644 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC008366 | |
DDC | |
DDC (AAH08366, 1 a.a. ∼ 480 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.