Learn More
DDX11, Mouse, Clone: 3C1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant DDX11.
Supplier: Abnova Corporation H00001663M03
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an enzyme that possesses both ATPase and DNA helicase activities. This gene is a homolog of the yeast CHL1 gene, and may function to maintain chromosome transmission fidelity and genome stability. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq
Sequence: MANETQKVGAIHFPFPFTPYSIQEDFMAELYRVLEAGKIGIFESPTGTGKSLSLICGALSWLRDFEQKKREEEARLLETGTGPLHDEKDESLCLSSSCEGAAGTPRPAGEPAWVTQFVQKKEERDLVDRLKAEQARRKQREERLQQLQHRVQLKYAAKRLRQEEEERENLLRLSREMLETGPEAERLEQLESGEEELVLAEYESDEEKKVASRVDEDEDDLEEEHITKIYYCSRTHSQLAQFVHEVKKSPFGKDVRLVSLGSRQNLCVNEDVKSLGSVQLINDRCVDMQRSRHEKKKGAEEEKPKRRRQEKQAACPFYNHEQMGLLRDEALAEVKDMEQLLALGKEARACPYYGSRLAIPAAQLVVLPYQMLLHAATRQAAGIRLQDQVVIIDEAHNLIDTITGMHSVEVSGSQLCQAHSQLLQYVERYGKRLKAKNLMYLKQILYLLEKFVAVLGGNIKQNPNTQSLSQTGTELKTINDFLFQSQIDNINLFKVQRYCEKSMISRKLFGFTERYGAVFSSREQPKLAGFQQFLQSLQPRTTEALAAPADESQASTLRPASPLMHIQGFLAALTTANQDGRVILSRQGSLSQSTLKFLLLNPAVHFAQVVKECRAVVIAGGTMQPVSDFRQQLLACAGVEAERVVEFSCGHVIPPDNILPLVICSGISNQPLEFTFQKRELPQMMDEVGRILCNLCGVVPGGVVCFFPSYEYLRQVHAHWEKGGLLGRLAARKKIFQEPKSAHQVEQVLLAYSRCIQACGQERGQVTGALLLSVVGGKMSEGINFSDNLGRCVVMVGMPFPNIRSAELQEKMAYLDQTLSPRPGTPREGSGGEPVHEGRQPVHRQGHQAPEGFCQRSAPGPAICPAPCPGQAAGLDPSPCGGQSYLWPRHCCCAEVSPGEVGLFLMGNHATAWRRALPLSCPLETVFVVGVVCGDPVTKVKPRRRVWSPECCQDPGTGVSSRRRKWGNPESpecifications
DDX11 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011264 | |
DDX11 | |
DDX11 (AAH11264, 1 a.a. ∼ 970 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
3C1 | |
Mouse monoclonal antibody raised against a full-length recombinant DDX11. | |
DDX11 | |
CHL1/CHLR1/KRG2/MGC133249/MGC9335 | |
Mouse | |
Affinity chromatography | |
RUO | |
1663 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.