Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016765 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-765 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-765 Supplier Abnova Corporation Supplier No. H00001663B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DDX11 protein.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an enzyme that possesses both ATPase and DNA helicase activities. This gene is a homolog of the yeast CHL1 gene, and may function to maintain chromosome transmission fidelity and genome stability. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: MANETQKVGAIHFPFPFTPYSIQEDFMAELYRVLEAGKIGIFESPTGTGKSLSLICGALSWLRDFEQKKREEEARLLETGTGPLHDEKDESLCLSSSCEGAAGTPRPAGEPAWVTQFVQKKEERDLVDRLKAEQARRKQREERLQQLQHRVQLKYAAKRLRQEEEERENLLRLSREMLETGPEAERLEQLESGEEELVLAEYESDEEKKVASRVDEDEDDLEEEHITKIYYCSRTHSQLAQFVHEVKKSPFGKDVRLVSLGSRQNLCVNEDVKSLGSVQLINDRCVDMQRSRHEKKKGAEEEKPKRRRQEKQAACPFYNHEQMGLLRDEALAEVKDMEQLLALGKEARACPYYGSRLAIPAAQLVVLPYQMLLHAATRQAAGIRLQDQVVIIDEAHNLIDTITGMHSVEVSGSQLCQAHSQLLQYVERYGKRLKAKNLMYLKQILYLLEKFVAVLGGNIKQNPNTQSLSQTGTELKTINDFLFQSQIDNINLFKVQRYCEKSMISRKLFGFTERYGAVFSSREQPKLAGFQQFLQSLQPRTTEALAAPADESQASTLRPASPLMHIQGFLAALTMANQDGRVILSRQGSLSQSTLKFLLLNPAVHFAQVVKECRAVVIAGGTMQPVSDFRQQLLACAGVEAERVVEFSCGHVIPPDNILPLVICSGISNQPLEFTFQKRELPQMMDEVGRILCNLCGVVPGGVVCFFPSYEYLRQVHAHWEKGGLLGRLAARKKIFQEPKSAHQVEQVLLAYSRCIQACGQERGQVTGALLLSVVGGKMSEGINFSDNLGRCVVMVGMPFPNIRSAELQEKMAYLDQTLSPRPGTPREGSGGEPVHEGRQPVHRQGHQAPEGFCQRSAPGPAICPAPCPGQAAGLDPSPCGGQSYLWPRHCCCAEVSPGEVGLFLMGNHTTAWRRALPLSCPLETVFVVGVVCGDPVTKVKPRRRVWSPECCQDPGTGVSSRRRKWGNPE

Specifications

Antigen DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DDX11 protein.
Formulation PBS with no preservative; pH 7.4
Gene DDX11
Gene Accession No. BC050522
Gene Alias CHL1/CHLR1/KRG2/MGC133249/MGC9335
Gene Symbols DDX11
Host Species Mouse
Immunogen DDX11 (AAH50522.1, 1 a.a. ∼ 970 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1663
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.