Learn More
DDX17, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human DDX17 peptide using ARM Technology.
Supplier: Abnova Corporation H00010521K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG (CUG) translation initiation codon, have been described for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
DDX17 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
DDX17 | |
DDX17 | |
A synthetic peptide of human DDX17 | |
RUO | |
10521 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
DEAD (Asp-Glu-Ala-Asp) box polypeptide 17 | |
DDX17 | |
DKFZp761H2016, P72, RH70 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.