Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DDX19A/B Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10068149
Description
DDX19A/B Polyclonal specifically detects DDX19A/B in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
DDX19A/B | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DDX19A DEAD (Asp-Glu-Ala-Asp) box polypeptide 19A DDX19-DDX19L, DDX19L | |
Goat | |
Affinity Purified | |
RUO | |
55308 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:4000 | |
NP_009173.1 | |
DDX19A | |
Peptide with sequence C-DLPVDKDGNPDNETY corresponding to internal region according to NP_001014451.1, NP_001014449.1, NP_060802.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize DDX19A (NP_060802.1) and all reported isoforms of DDX19B (NP_009173.1, NP_001014451.1 and NP_001014449.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction