Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006609 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-609 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-609 Supplier Abnova Corporation Supplier No. H00008653B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DDX3Y protein.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, and it has a homolog on the X chromosome. The gene mutation causes male infertility, Sertoli cell-only syndrome or severe hypospermatogenesis, suggesting that this gene plays a key role in the spermatogenic process. Alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Sequence: MSHVVVKNDPELDQQLANLDLNSEKQSGGASTASKGRYIPPHLRNREASKGFHDKDSSGWSCSKDKDAYSSFGSRDSRGKPGYFSERGSGSRGRFDDRGRSDYDGIGNRERPGFGRFERSGHSRWCDKSVEDDWSKPLPPSERLEQELFSGGNTGINFEKYDDIPVEATGSNCPPHIENFSDIDMGEIIMGNIELTRYTRPTPVQKHAIPIIKGKRDLMACAQTGSGKTAAFLLPILSQIYTDGPGEALKAVKENGRYGRRKQYPISLVLAPTRELAVQIYEEARKFSYRSRVRPCVVYGGADIGQQIRDLERGCHLLVATPGRLVDMMERGKIGLDFCKYLVLDEADRMLDMGFEPQIRRIVEQDTMPPKGVRHTMMFSATFPKEIQMLARDFLDEYIFLAVGRVGSTSENITQKVVWVEDLDKRSFLLDILGATGSDSLTLVFVETKKGADSLEDFLYHEGYACTSIHGDRSQRDREEALHQFRSGKSPILVATAVAARGLDISNVRHVINFDLPSDIEEYVHRIGRTGRVGNLGLATSFFNEKNMNITKDLLDLLVEAKQEVPSWLENMAYEHHYKGGSRGRSKSNRFSGGFGARDYRQSSGSSSSGFGASRGSSSRSGGGGYGNSRGFGGGGYGGFYNSDGYGGNYNSQGVDWWGN

Specifications

Antigen DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DDX3Y protein.
Formulation No additive
Gene DDX3Y
Gene Accession No. NM_004660.2
Gene Alias DBY
Gene Symbols DDX3Y
Host Species Mouse
Immunogen DDX3Y (NP_004651.2, 1 a.a. ∼ 660 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8653
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.