Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEAD (Asp-Glu-Ala-Asp) box polypeptide 50, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008146 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-146 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-146 Supplier Abnova Corporation Supplier No. H00079009B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human DDX50 protein.

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. This gene and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. This gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined. [provided by RefSeq

Sequence: MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFD

Specifications

Antigen DEAD (Asp-Glu-Ala-Asp) box polypeptide 50
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human DDX50 protein.
Formulation No additive
Gene DDX50
Gene Accession No. NM_024045.1
Gene Alias GU2/GUB/MGC3199/RH-II/GuB
Gene Symbols DDX50
Host Species Mouse
Immunogen DDX50 (NP_076950.1, 1 a.a. ∼ 737 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 79009
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.