Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Dectin-1/CLEC7A, Alexa Fluor 700, Clone: 259931, R&D Systems™

Catalog No. FAB1859N100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1859N100 100 Tests
FAB1859N025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1859N100 Supplier R&D Systems Supplier No. FAB1859N100
This item is not returnable. View return policy

Specifications

Antigen Dectin-1/CLEC7A
Applications Flow Cytometry
Classification Monoclonal
Clone 259931
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_072092
Gene Alias Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Gene Symbols CLEC7A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Dectin-1/CLEC7A, Thr66-Met201, Accession # NP_072092
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64581
Test Specificity Detects human Dectin-1/CLEC7A in direct ELISAs. In directs ELISAs, less than 10% cross-reactivity with recombinant human (rh) DLEC and no cross-reactivity with recombinant mouse Dectin-1 or rhDectin-2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.