Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Defensin Β3 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3478MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3478MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3478MI Supplier MilliporeSigma Supplier No. AB3478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Defensin Antibody, β3 is an antibody against Defensin for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Defensin β3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols DEFB103A; HBD3; HBD-3; BD-3; HBP-3; hBD-3; BD3; DEFB103; DEFB3; DEFB103B; HBP3; DEFB-3
Host Species Rabbit
Immunogen A synthetic peptide consisting of amino acids 23-33 [GIINTLQKYYC] of the human beta-defensin-3 protein.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_018661.3
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.