Learn More
Defensin, clone: DEF-3, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA182491
Description
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. It differs from defensin, alpha 3 by only one amino acid.
This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. A suggested positive control for immunohistochemical applications is human tonsil.Reconstitute with 0.5 ml of distilled water. After reconstitution, store product undiluted at 4ºC or -20ºC if preferred.
Specifications
Defensin | |
Monoclonal | |
0.2mg/mL | |
DEFA1 | |
DEF1, DEFA2, HNP-1, HP-1, MGC138393, MRS | |
Mouse | |
Purified | |
RUO | |
1667, 1668, 1669, 1670 | |
4° C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
DEF-3 | |
Unconjugated | |
P12838, P59665, P59666, Q01523 | |
DEFA1, DEFA3, DEFA4, DEFA5 | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.