Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEGS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159941
Description
DEGS1 Polyclonal specifically detects DEGS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DEGS1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Cell migration-inducing gene 15 protein, Degenerative spermatocyte homolog 1, degenerative spermatocyte homolog 1, lipid desaturase (Drosophila), degenerative spermatocyte homolog, lipid desaturase (Drosophila), DEGS, Des-1, DES1degenerative spermatocyte homolog, lipid desaturase, EC 1.14, FADS7, membrane fatty acid (lipid) desaturase, Membrane lipid desaturase, MGC5079, migration-inducing gene 15 protein, MLDdihydroceramide desaturase, sphingolipid delta 4 desaturase, sphingolipid delta(4)-desaturase DES1 | |
Rabbit | |
38 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Rat: 92%; Sheep: 92%; Zebrafish: 92%; Canine: 83%; Chicken: 78%. | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
O15121 | |
DEGS1 | |
Synthetic peptides corresponding to DEGS1(degenerative spermatocyte homolog 1, lipid desaturase (Drosophila)) The peptide sequence was selected from the N terminal of DEGS1 (NP_003667). Peptide sequence GSRVSREDFEWVYTDQPHADRRREILAKYPEIKSLMKPDPNLIWIIIMMV The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
8560 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction