Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEK Mouse anti-Human, Unlabeled, Clone: 2, BD

Catalog No. BDB610948 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610948 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610948 Supplier BD Biosciences Supplier No. 610948
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The (6;9) chromosomal translocation is associated with acute myelogenousleukemia (AML) and fuses the dek and can genes. This results in expression of the oncogenic DEK-CAN fusion protein, consisting of the N-terminal two-thirds of DEK and the C-terminal two-thirds of CAN. Although, on its own, DEK exhibits anti-oncogenic properties, the DEK-CAN chimera appears to be oncogenic. DEK is a nuclear protein with a calculated molecular weight of 42-43 kD, that can be observable at 50 kD, and reportedly exhibits no substantial homology to any known protein sequences. Although it contains 42% charged amino acids and multiple acidic sequences, specific structural features have yet to be identified. In addition to its involvement in AML, DEK is associated with several disease states, such as juvenile rheumatoid arthritis where it is an autoantigen. Efforts to define the cellular function of DEK led to its identification as the pets factor. The peri-ets (pets) site is a TG-rich element between the two Elf-1 binding sites of the HIV-2 enhancer. The pets site mediates transcriptional activation in response to T cell stimulation. Thus, DEK is a site-specific DNA binding protein that functions in transcriptional regulation and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEK
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human DEK aa. 19-169
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.