Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDIL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522998
Description
Suggested positive control: normal human brain lysate, normal mouse brain lysate.
Del-1 is an integrin ligand that is expressed by endothelial cells during embryological vascular development. It promotes adhesion of endothelial cells through an interaction with the alpha-v/beta-3 integrin receptor. Del-1 plays a vital role in mediating angiogenesis.
Specifications
EDIL3 | |
Polyclonal | |
Unconjugated | |
EDIL3 | |
Del1; Del-1; developmental endothelial locus-1; developmentally-regulated endothelial cell locus 1 protein; Edil3; EGF like repeats and discoidin domains 3; EGF-like repeat and discoidin I-like domain-containing protein 3; EGF-like repeats and discoidin I-like domains 3; integrin-binding protein DEL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10085, 13612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1.62 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
O35474, O43854 | |
EDIL3 | |
Synthetic peptide made to an internal portion of the human protein (within residues 200-350). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction