Learn More
Delta-Like Protein 1 Mouse anti-Human, PE, Clone: MHD1-314, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564414
Description
The MHD1-314 monoclonal antibody specifically binds to Delta-Like Protein 1 which is also known as Delta1, DLL1, and DL1. Delta-Like Protein 1 is a type 1 transmembrane glycoprotein which belongs to the Delta/Serrate/Jagged Family of Notch receptor ligands. The Notch family of transmembrane receptors and their ligands play key roles in various developmental and postnatal processes by controlling cell-fate decisions. Delta-Like Protein 1 is expressed by cells within heart, pancreas, brain, muscle and hematopoietic/lymphoid tissues. This Notch ligand functions in the development and postnatal activities of the nervous and skeletal systems. It also functions in the hematopoietic and immune systems, for example, by promoting the differentiation of hematopoietic progenitors into T cell and NK cell precursors. The MHD1-314 antibody reportedly inhibits the development of osteoclasts from cultured peripheral blood monocytes suggesting that Delta-Like Protein 1 positively regulates osteoclastogenesis.

Specifications
Delta-Like Protein 1 | |
Monoclonal | |
0.2mg/mL | |
DLL1; DL1; Delta1; H-Delta-1; Delta-like 1; Delta | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
MHD1-314 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human DLL1 Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.