Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta-Like Protein 4 Mouse anti-Human, PE, Clone: MHD4-46, BD

Catalog No. BDB564412 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564412 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564412 Supplier BD Biosciences Supplier No. 564412
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MHD4-46 monoclonal antibody specifically binds to Delta-Like Protein 4 which is also known as Delta4, DLL4, and DL4. Delta-Like Protein 4 is a type 1 transmembrane glycoprotein which belongs to the Delta/Serrate/Jagged Family of Notch receptor ligands. The Notch family of transmembrane receptors and their ligands play key roles in various developmental and postnatal processes by controlling cell-fate decisions. Delta-Like Protein 4 is expressed by vascular endothelial cells within certain normal tissues and tumors. It signals through Notch1 and Notch 4 Receptors and plays a critical role in vascular development and homeostasis. Delta-Like Protein 4 expression is upregulated in response to vascular endothelial growth factor (VEGF) and regulates endothelial cell proliferation during vascular sprouting. Delta-Like Protein 4 is also expressed by activated human monocytes and dendritic cells that have been stimulated with proinflammatory activators such as lipopolysaccharide. Recent evidence suggests that Delta-Like Protein 4 may contribute to the T cell potential of CD34+ cells and to the growth of various tumors, and promote tumor cell infiltration.

During product development, the specificity of the purified MHD4-46 antibody (Cat. No. 564589) was confirmed by immunohistochemical staining of acetone-fixed frozen sections of human placenta. Vascular endothelial cells were specifically stained (data not shown).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta-Like Protein 4
Applications Flow Cytometry
Classification Monoclonal
Clone MHD4-46
Concentration 0.2mg/mL
Conjugate PE
Description DLL4; Delta-like 4; DL4; Delta ligand 4; Delta4; Notch ligand DLL4
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Delta-Like Protein 4 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.