Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Delta3 Mouse anti-Human, Clone: 1E7.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT189
Description
This Anti-Delta3 Antibody, clone 1E7.2 is validated for use in western blot, FC for the detection of Delta3.
Specifications
Delta3 | |
Monoclonal | |
Unconjugated | |
NP_058637 | |
Mouse | |
Purified by ion exchange chromatography | |
Primary | |
2-8°C for one year | |
IgM κ |
Flow Cytometry, Western Blot | |
1E7.2 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
DLL3 | |
Linear peptide corresponding to human Delta3. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction