Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-deltaNp73 Clone: 38C674,

Catalog No. OP181100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP181100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP181100UG Supplier MilliporeSigma™ Supplier No. OP181100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ΔNp73 Clone: 38C674 in Human samples, and it is validated for Immuno Blotting

p73 is a relative of tumor suppressor protein p53. It displays a high homology with p53 and with p63, a gene implicated in the maintenance of epithelial stem cells. Significant homology between p53, p63, and p73 suggests that they may have overlapping functions involved in the regulation of gene expression. The disruption of p73 leads to hippocampal dysgenesis, hydrocephalus, chronic inflammation, and infections. Splicing variant mRNAs of p73 have been identified in MCF7, a breast carcinoma cell line. These mRNAs code for variant p73 proteins bearing distinct carboxyl-terminal structures, suggesting that this region of p73 may be important for the functions of this protein. Researchers have recently cloned a p73 variant that lacks an amino-terminal transactivation domain, called ΔNp73. The ΔNp73 message is transcribed from an alternative promoter located in intron 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ΔNp73
Applications Immunoblot
Classification Monoclonal
Clone 38C674
Conjugate Unconjugated
Formulation In PBS, 0.2% gelatin.
Host Species Mouse
Immunogen a synthetic peptide (LYVGDPARHLAT) corresponding to amino acids 2-13 of human ΔNp73
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽73kDa ΔNp73 protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.