Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DENND5A Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524015
Description
This antibody is predicted to react with mouse based on sequence homology.
May be involved in Rab6-mediated GTPase signaling.
Specifications
| DENND5A | |
| Polyclonal | |
| Unconjugated | |
| Q6IQ26 | |
| DENND5A | |
| KLH conjugated synthetic peptide between 695-724 amino acids from the central region of human DEN5A | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| DENND5A | |
| RAB6IP1, DENN domain-containing protein 5A, RAB6 interacting protein 1, rab6-interacting protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23258 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction