Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dendritic Cells Mouse anti-Rat, Unlabeled, Clone: OX-62, BD

Catalog No. BDB555010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555010 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555010 Supplier BD Biosciences Supplier No. 555010

Mouse Monoclonal Antibody

The OX-62 antibody reacts with an antigen on dendritic cells, dendritic epidermal T cells (but not RT1B+ Langerhans cells), and intestinal intraepithelial lymphocytes of normal rats and on CD3+ lymphocytes in the spleen and cervical lymph nodes of athymic nude rats. The antigen can be detected by flow cytometric or immunocytochemical analysis of leukocytes and by immunohistochemical staining of epidermal sheets, wholemounts of neural tissue, and frozen sections of lymphoid and non-lymphoid organs. The OX-62 mAb immunoprecipitates a molecule with integrin-like properties, and sequencing indicates that it is related to mouse and human CD103 (Integrin αIEL or αE chain).

Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Dendritic Cells
Applications Flow Cytometry
Classification Monoclonal
Clone OX-62
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Density gradient-enriched PVG rat thoracic-duct dendritic cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.