Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desert hedgehog protein (DHH) Mouse anti-Rat, Human, Clone: 19D7.2, MilliporeSigma™

Catalog No. 04967MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-967-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-967-MI Supplier MilliporeSigma Supplier No. 04967
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Desert hedgehog protein (DHH) using this Anti-Desert hedgehog protein (DHH) Antibody, clone 19D7.2 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desert hedgehog protein (DHH)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 19D7.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_066382
Gene Symbols DHH, HHG-3
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Desert hedgehog protein.
Purification Method Ion Exchange
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.