Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Mouse anti-Human, Pig, Rat, Frog, Mouse, Clone: DE-B-5, MilliporeSigma™

Catalog No. MAB3430MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3430MI 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3430MI Supplier MilliporeSigma Supplier No. MAB3430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Desmin using this Anti-Desmin Antibody, clone DE-B-5 validated for use in western blot, immunohistochemistry, immunohistochemistry (paraffin), immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DE-B-5
Conjugate Unconjugated
Formulation Liquid in 0.02Mphosphate buffer, 0.25M NaCl with 0.1% sodium azide, pH 7.6.
Gene Accession No. NM_001927.3
Gene Symbols DES, CSM2, FLJ41013, FLJ39719, CSM1, FLJ41793, FLJ12025, Desmin., CMD1I, desmin
Host Species Mouse
Immunogen Purified desmin.
Quantity 40 μg
Primary or Secondary Primary
Target Species Frog, Human, Mouse, Pig, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.