Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin Rabbit anti-Bovine, Chicken, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB907MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB907MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB907MI Supplier MilliporeSigma Supplier No. AB907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Desmin Antibody is an antibody against Desmin for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Desmin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum. Liquid containing 0.1% sodium azide as a preservative.
Gene Symbols DES; CSM2; FLJ41013; FLJ39719; CSM1; FLJ41793; FLJ12025; Desmin.; CMD1I; desmin
Host Species Rabbit
Immunogen Purified desmin from chicken gizzard, isolated by modified procedure of Geisler, et al [J. Biol Chem. (1980). 111:425-433].
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001927.3
Target Species Bovine, Chicken, Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.