Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFF45/35 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB16961MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB16961MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB16961MI Supplier MilliporeSigma Supplier No. AB16961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-DFF45/35 Antibody, N-terminus is validated for use in immunoprecipitation, western blotting for the detection of DFF45/35.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DFF45/35
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols DFFA; DFF45; DFF-45; ICAD; DFF1
Host Species Rabbit
Immunogen Polyclonal antibody raised against a peptide corresponding to amino acids 2-21 at N-terminus of human DFF (Liu et al. 1997).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004401.2; NM_213566.1
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.