Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGAT2, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203688 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-688 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-688 Supplier Abnova Corporation Supplier No. H00084649K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human DGAT2 peptide using ARM Technology.

Acyl-CoA:diacylglycerol acyltransferase, or DGAT (EC 2.3.1.20), is responsible for the synthesis of triglycerides. It catalyzes a reaction in which diacylglycerol is covalently joined to long chain fatty acyl-CoAs.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen DGAT2
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description diacylglycerol O-acyltransferase homolog 2 (mouse)
Formulation In 1X PBS, pH 7.4
Gene DGAT2
Gene Accession No. DGAT2
Gene Alias DKFZp686A15125, HMFN1045
Gene Symbols DGAT2
Host Species Rabbit
Immunogen A synthetic peptide of human DGAT2
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84649
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.